Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) sulfamate, 50% w/w aq. soln., Reagent Grade

Catalog No. AA405018M
Click to view available options
Quantity:
1 L
2 x 2.5 L

Catalog No. AA405018M

Supplier: Thermo Scientific Chemicals 0405018M

Only null left
Add to Cart

Nickel plating | CAS: 13770-89-3 | H4N2NiO6S2 | 250.85 g/mol

Nickel plating Nickel(II) sulfamate is used for electroplating and as a surface treating agent. It finds application in metal coloring and casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13770-89-3
Molecular Formula H4N2NiO6S2
Molecular Weight (g/mol) 250.85
MDL Number MFCD00137261
InChI Key KERTUBUCQCSNJU-UHFFFAOYSA-L
Synonym nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci
PubChem CID 83720
IUPAC Name nickel(2+);disulfamate
SMILES [Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O

Specifications

Color Green
Physical Form Liquid
Quantity 2 x 2.5 L
Linear Formula Ni(SO3NH2)2
UN Number UN3082
Solubility Information Miscible with water,immiscible with ether,acetone.
Formula Weight 250.86 (Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Nickel(II) sulfamate
Show More Show Less
Hazard Category H317-H334-H341-H350i-H360D-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 237-396-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.