Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Nickel (II) Sulfate, Hexahydrate
SCI_ED

Catalog No. S25445 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25445 100 g Reagent Grade
S25445A 500 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25445

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Color Blue-Green
Quantity 100 g
Assay Percent Range >95%
Linear Formula NiSO4·6H2O
Formula Weight 262.87
Grade Reagent
Packaging Reagent Grade
Chemical Name or Material Nickel (II) Sulfate, Hexahydrate
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 6.1, III, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.