Learn More
Nickel(II) sulfate hexahydrate, 98%
Description
Nickel sulfate hexahydrate is mainly used in Ni plating, as mordant in dyeing and in printing textiles, as a blackening agent for zinc and brass and in the fabrication of organic nickel salts. It is used as an effective catalyst for gas-phase dehydration of glycerol to acrolein. Nickel sulfate hexahydrate crystal is an efficient ultraviolet filter material. Columns used in biochemistry and molecular biology can be regenerated with nickel sulfate. It is used to prepare nickel carbonate, a precursor to pigments and nickel-based catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| pH | 4.3 to 4.7 |
| Physical Form | Crystalline |
| Quantity | 2 kg |
| Assay Percent Range | 98% |
| Linear Formula | NiSO4·6H2O |
| UN Number | UN3288 |
| Merck Index | 14,6517 |
| Solubility Information | Very soluble in water,ethanol,ammonia. |
| Formula Weight | 262.86 (154.77 Anhydrous) |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H317-H319-H332-H334-H335-H341-H350i-H360D-H372 |
| Hazard Statement | GHS H Statement H334-H350-H360-H372-H341-H302-H332-H315-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Harmful if swallowed. Harmful if inhaled. Causes skin irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 232-104-9 |
| RTECSNumber | QR9600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.