Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydroxide, ca. 85%, ACS reagent, pellets

Catalog No. AC424140251
Click to view available options
Quantity:
25 g
500 g
2.5 kg
5 kg
25 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1310-58-3 | HKO | 56.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color White
Boiling Point 1320.0°C
Melting Point 360.0°C
Assay Percent Range ca. 85%
Linear Formula KOH
Fieser 01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258
Merck Index 15, 7761
Solubility Information Solubility in water: 1120g/L (20°C). Other solubilities: 1780g/L water (100°C), soluble in ethanol (400g/L), soluble in ammonia, slightly soluble in ether
Quantity 25 kg
Formula Weight 56.11
Percent Purity ∼85%
Grade ACS Reagent
Packaging Fibre drum
Physical Form Pellet
Chemical Name or Material Potassium hydroxide, 85%
Show More Show Less
Hazard Category Danger
EINECSNumber 215-181-3
RTECSNumber TT2100000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.