Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel (II) Sulfate Hexahydrate, Purified, Reagents

Catalog No. 01337156
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-156 500 g
01-337-157 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-156 Supplier Reagents Holdings Llc Supplier No. C2256500500A5
Add to Cart
Add to Cart

Nickel (II) Sulfate Hexahydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.84
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym Nickelous Sulfate Hexahydrate, Nickel (II) Sulfate Hexahydrate
IUPAC Name nickel(2+) hexahydrate sulfate
SMILES O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 103°C
Color Blue to Green
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 3.68
Chemical Name or Material Nickel (II) Sulfate Hexahydrate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.