Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) telluride, 99.9% (metals basis excluding Co), Co 0.1-1%

Catalog No. p-7049168
Click to view available options
Quantity:
10 g
50 g

CAS: 12142-88-0 | NiTe | 186.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12142-88-0
Molecular Formula NiTe
Molecular Weight (g/mol) 186.29
MDL Number MFCD00016258
InChI Key NPEUSMKUOOTUGX-UHFFFAOYSA-N
Synonym nickel telluride, imgreite, ccris 7583, nickel telluride nickel and nickel compounds, nickeltellurid
PubChem CID 62780
IUPAC Name tellanylidenenickel
SMILES [Ni]=[Te]

Specifications

Physical Form Lump
Quantity 10 g
Linear Formula NiTe
UN Number UN3077
Formula Weight 186.31
Percent Purity 99.9%
Odor Odorless
Assay (metals basis excluding Co), Co 0.1 to 1%
Chemical Name or Material Nickel(II) telluride
Hazard Category H317-H350i-H372
Hazard Statement GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 235-260-6
RTECSNumber QS0400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.