Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel molybdenum oxide, 98%

Catalog No. p-7080515
Click to view available options
Quantity:
25 g
100 g

Catalog No. AA8993814

Supplier: Thermo Scientific Chemicals 08993814

Only null left
Add to Cart

CAS: 14177-55-0 | MoNiO4 | 218.639 g/mol

Nickel molybdenum oxide is used as catalyst in petroleum industry for the oil fraction like naptha, kerosene and diesel. It is also used extensively as a chemical intermediate for catalytic purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14177-55-0
Molecular Formula MoNiO4
Molecular Weight (g/mol) 218.639
MDL Number MFCD00016255
InChI Key NLPVCCRZRNXTLT-UHFFFAOYSA-N
Synonym molybdenum nickel tetraoxide, nickel molybdate, nickel 2+ molybdate, molybdenum nickel oxide, nickel 2+ dioxido dioxo molybdenum, moo4.ni, nickel molybdenum oxide, dioxido dioxo molybdenum; nickel 2+
PubChem CID 84241
IUPAC Name dioxido(dioxo)molybdenum;nickel(2+)
SMILES [O-][Mo](=O)(=O)[O-].[Ni+2]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Soluble in water (4652mg/L at 20C).
Formula Weight 218.65
Percent Purity 98%
Odor Odorless
Chemical Name or Material Nickel molybdenum oxide
Hazard Category H317-H350i-H372
Hazard Statement GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 238-034-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.