Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nickel Monoclonal Antibody (4E3)
GREENER_CHOICE

Catalog No. PIMA549670
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549670 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549670 Supplier Invitrogen™ Supplier No. MA549670
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Nickel is a chemical element with the symbol Ni and atomic number 28. It is a silvery-white lustrous metal with a slight golden tinge. Nickel is one of the four elements that are ferromagnetic at room temperature, along with iron, cobalt, and gadolinium. It is a hard, ductile metal that resists corrosion and tarnishing. Nickel is widely used in the production of stainless steel, as well as in alloys for various applications including coinage, electronics, and aerospace. It is also an important component of batteries, catalysts, and pigments. Nickel is found in the Earth's crust, but it is mainly extracted from nickel sulfide and nickel laterite ores.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nickel
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 4E3
Concentration 2.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of Ni hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.