Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL

Catalog No. AA13839AD
Click to view available options
Quantity:
50 mL
100 mL
500 mL

Catalog No. AA13839AD

Supplier: Thermo Scientific Chemicals 013839AD

Only null left
Add to Cart

CAS: 7440-02-0 | Ni | 58.69 g/mol

Nickel, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Molecular Formula Ni in 5% HNO3
Concentration Ni 1000 μg/mL
MDL Number MFCD00011137
UN Number UN3264
Solubility Miscible with water
Quantity 50 mL
Chemical Name or Material Nickel, plasma standard solution, Specpure™
Hazard Category H290-H314-H335-H350i-H360D
Hazard Statement GHS H Statement
H314-H317-H208
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.