Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
Description
Conductive surface coatings, inks, adhesives, and sealants
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Silver to Gray |
| Appearance | Silver gray powder |
| Density | 8.908g/cm3 at 20°C |
| Quantity | 100 g |
| Mesh Size | -325 Mesh |
| Odor | Odorless |
| Form | Conductive Dendritic Powder |
| Assay Percent Range | 99.8% (Metals basis) |
| Chemical Name or Material | Nickel |
Safety and Handling
| Hazard Category | H228-H317-H350-H372 |
| Hazard Statement | Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets. |
| Precautionary Statement | P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 4.1, Packaging Group: II |
| EINECSNumber | 231-111-4 |
| RTECSNumber | QR5950000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction