Learn More
Nickel on silica-alumina, catalyst
Description
Nickel on Silica-Alumina Catalys products are often used as the most efficient way to catalyze organic reactions. It is also used in R&D, manufacture of chemicals, Pharmaceuticals and intermediates. It is an important raw material and intermediate used in Organic Synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Black |
Appearance | Black catalyst powder |
Quantity | 25 g |
Odor | Odorless |
Form | Powder |
Solubility Information | It is insoluble in water. |
Chemical Name or Material | Nickel, on silica-alumina, catalyst |
Safety and Handling
Hazard Category | H228-H317-H350i-H372 |
Precautionary Statement | P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c |
DOTInformation | DOT Class: 4.1, Packaging Group: II |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.