Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sponge Nickel, A-4000, promoted with 2% Iron and 2.5% Chromium, Thermo Scientific Chemicals

Catalog No. p-4782160
Click to view available options
Quantity:
100 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Gray
Appearance Dark gray sponge
Quantity 100 g
Sensitivity Air Sensitive
Form Powder
Chemical Name or Material Nickel
Hazard Category H250-H317-H350-H372-X
Hazard Statement Wear suitable protective clothing and gloves.
Precautionary Statement P201-P202-P210-P222-P235+P410-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P335+P334-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.2, Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.