Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel sulfide, 99.9% (metals basis)

Catalog No. p-4407665
Click to view available options
Quantity:
10 g
50 g

CAS: 12035-72-2 | Ni3S2 | 240.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12035-72-2
Molecular Formula Ni3S2
Molecular Weight (g/mol) 240.20
MDL Number MFCD01711880
InChI Key YGHCWPXPAHSSNA-UHFFFAOYSA-N
Synonym nickel subsulfide, trinickel disulphide, nickel sulfide, trinickel disulfide, nickel subsulphide, nickel sesquisulfide, nickel tritadisulphide, nickel iii sulfide, alpha nickel subsulfide, ccris 432
PubChem CID 22833473
SMILES S=[Ni][Ni][Ni]=S

Specifications

Melting Point 797°C
Color Black
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water(0,00735g/L at 20°C).
IUPAC Name disulfanylidenetrinickel
Formula Weight 240.19
Appearance Black powder
Chemical Name or Material Nickel sulfide
Hazard Category H317-H341-H350i-H372
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation DOT Class: 9; Packing Group: III
EINECSNumber 234-829-6
RTECSNumber QR9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.