Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel sulfide, 99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7059862
Click to view available options
Quantity:
5 g

Nickel sulfide is used as a semiconductor material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12035-51-7
Molecular Formula NiS2
Molecular Weight (g/mol) 122.81
MDL Number MFCD00016267
InChI Key HNDMNBUITRNJKK-UHFFFAOYSA-N
Synonym nickel iv sulphide
PubChem CID 131675689
IUPAC Name disulfanylidenenickel
SMILES S=[Ni]=S

Specifications

Physical Form Powder
Quantity 5 g
UN Number UN3077
Solubility Information Insoluble in water.
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Nickel sulfide
Hazard Category H317-H341-H350-H372
Hazard Statement GHS H Statement
H350-H372-H341-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.