Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NICN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566124
Description
Immunogen sequence: PSVTFPKWLSH PVPCEQPALL REGLPDPSRV SSEVQQMWAL TEMIRASHTS ARIGRFDVDG CYDLNLLSYT Highest antigen sequence identity to the following orthologs - mouse 89%, rat 87%.
This protein encoded by this gene localizes to the nucleus and is expressed in numerous tissues including brain, testis, liver, and kidney. Computer predictions indicate that this region of the 3' UTR contains hairpin-forming self-complementary sequence which is possibly excised after transcription. This gene has a pseudogene on chromosome X.
Specifications
NICN1 | |
Polyclonal | |
Unconjugated | |
NICN1 | |
NICN1; nicolin 1; nicolin-1; NPCEDRG; PGs5; Tubulin polyglutamylase complex subunit 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84276 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BSH3 | |
NICN1 | |
Recombinant Human NICN1. Recombinant protein control fragment (Product #RP-105090). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction