Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinamide, 99%

Catalog No. AAA159700B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA159700B 1000 g
AAA1597030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA159700B Supplier Thermo Scientific Chemicals Supplier No. A159700B
Add to Cart

CAS: 98-92-0 | C6H6N2O | 122.127 g/mol

Nicotinamide is a building block (active component) of coenzymes nicotinamide adenine dinucleotide (NAD) and nicotinamide adenine dinucleotide phopshate (NADP). It may be used as a substrate of nicotinamide phosphoribosyltransferase (NAMPT, EC 2.4.2.12), the rate-limiting enzyme for NAD(+) biosynthesis and an important signaling molecule. It is an amide derivative of vitamin B3 and a PARP inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nicotinamide is a building block (active component) of coenzymes nicotinamide adenine dinucleotide (NAD) and nicotinamide adenine dinucleotide phopshate (NADP). It may be used as a substrate of nicotinamide phosphoribosyltransferase (NAMPT, EC 2.4.2.12), the rate-limiting enzyme for NAD(+) biosynthesis and an important signaling molecule. It is an amide derivative of vitamin B3 and a PARP inhibitor.

Solubility
Soluble in water, ethanol , glycerol, DMSO , methanol, butanol and chloroform. Slightly soluble in ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00006395
InChI Key DFPAKSUCGFBDDF-UHFFFAOYSA-N
Synonym nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide
PubChem CID 936
ChEBI CHEBI:17154
IUPAC Name pyridine-3-carboxamide
SMILES C1=CC(=CN=C1)C(=O)N

Specifications

Melting Point 128°C to 132°C
Density 1.4
Flash Point 182°C (360°F)
Odor Odorless
Assay Percent Range 99%
Quantity 1000 g
Beilstein 383619
Merck Index 14,6523
Solubility Information Soluble in water,ethanol ,glycerol,DMSO ,methanol,butanol and chloroform. Slightly soluble in ether.
Formula Weight 122.13
Percent Purity 99%
Chemical Name or Material Nicotinamide
Show More Show Less
Hazard Category H319-H500
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 202-713-4
RTECSNumber QS3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.