Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Nicotinamide Adenine Dinucleotide, MP Biomedicals™

Catalog No. mp21004991
Click to view available options
Quantity:
100 mg
500 mg
5 g
10 g

Acts as a major electron acceptor molecule in biological oxidations. β-Nicotinamide Adenine Dinucleotide, MP Biomedicals serves as a coenzyme of the dehydrogenases and acts as a hydrogen acceptor. β-NAD is one of the biologically active forms of

β-NAD occurs in living cells primarily in the oxidized state.

  • Serves as a coenzyme of hydride-transfer enzymes in dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols
  • Usually acts as a hydrogen acceptor, forming NADH, which then serves as a hydrogen donor in the respiratory chain
  • Used as a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins
  • Recently reported to be a P2Y11 agonist
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-84-9
Molecular Formula C21H27N7O14P2
Molecular Weight (g/mol) 663.43
MDL Number MFCD00150377
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
Synonym nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
PubChem CID 15938971
ChEBI CHEBI:57540
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Absorbance λ max (water): 259 ± 3nm
Melting Point 160°C (literature)
pH 2 to 4 (1% aq. soln.)
Quantity 100 mg
Solubility Information Soluble in water (200mg/mL)
Formula Weight 663.4 (anhydrous)
Percent Purity ≥98%
Physical Form Powder
Recommended Storage 0°C, desiccate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.