Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Nicotinamide Adenine Dinucleotide, Oxidized, Free Acid, 99%, MP Biomedicals™

Catalog No. p-4404987
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 53-84-9
Molecular Formula C21H27N7O14P2
Molecular Weight (g/mol) 663.43
MDL Number MFCD00150377
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
Synonym nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
PubChem CID 15938971
ChEBI CHEBI:57540
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Color White
Assay Percent Range 0.99
Quantity 1 g
Formula Weight 663.4 (Anhydrous)
Physical Form Powder
Chemical Name or Material β-Nicotinamide Adenine Dinucleotide, Oxidized, Free Acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.