Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinamide N-Methyltransferase/NNMT Antibody (AT11G11), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250581IR
Description
Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Nicotinamide N-Methyltransferase/NNMT | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NNMT | |
Recombinant human NNMT (1-264aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
AT11G11 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 2.1.1.1, nicotinamide N-methyltransferase | |
Mouse | |
Protein A purified | |
RUO | |
4837 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction