Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinamide N-Methyltransferase/NNMT Antibody (AT11G11), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250581PECY55
Description
Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human samples. It is validated for Flow Cytometry.Specifications
Nicotinamide N-Methyltransferase/NNMT | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
NNMT | |
Recombinant human NNMT (1-264aa) purified from E. coli | |
0.1 mL | |
4837.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
AT11G11 | |
Flow Cytometry | |
EC 2.1.1.1, nicotinamide N-methyltransferase | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction