Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinamide N-Methyltransferase/NNMT Antibody (OTI3D8), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272984APCCY7
Description
Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
Nicotinamide N-Methyltransferase/NNMT | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
NNMT | |
Full length recombinant protein of human Nicotinamide N-Methyltransferase/NNMT (NP_006160) produced in HEK293T cells. | |
0.1 mL | |
4837.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry, Immunoassay | |
OTI3D8 | |
Flow Cytometry, Knockout Validated | |
EC 2.1.1.1, nicotinamide N-methyltransferase | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction