Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinamide N-Methyltransferase/NNMT Antibody (OTI3D8), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272984APC
Description
Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
| Nicotinamide N-Methyltransferase/NNMT | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| NNMT | |
| Full length recombinant protein of human Nicotinamide N-Methyltransferase/NNMT (NP_006160) produced in HEK293T cells. | |
| 0.1 mL | |
| 4837.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunoassay | |
| OTI3D8 | |
| Flow Cytometry, Knockout Validated | |
| EC 2.1.1.1, nicotinamide N-methyltransferase | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction