Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinamide N-methyltransferase, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABE178825UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE178825UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE178825UG Supplier MilliporeSigma Supplier No. ABE178825UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Nicotinamide N-methyltransferase Antibody, Cat. No. ABE1788, is a highly specific rabbit polyclonal antibody that targets human Nicotinamide N-methyltransferase and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.

Nicotinamide N-methyltransferase (UniProt: P40261; also known as EC: 2.1.1.1) is encoded by the NNMT gene (Gene ID: 4837) in human. Nicotinamide N-methyltransferase is a monomeric, cytoplasmic enzyme that belong to the class I -like SAM-binding methyltransferase superfamily. It catalyzes the N-methylation of nicotinamide and other pyridines with S-Adenosyl-L-methionine (SAM)) serving as the methyl donor. It has two SAM binding sites (aa 63-64 and 142-143). Its catalytic activity is important for biotransformation of many drugs and xenobiotic compounds. Nicotinamide N-methyltransferase is predominantly expressed in the liver. A lower level of expression is seen in the kidney, lung, skeletal muscle, placenta, and heart and it is not detected in the brain or pancreas. Nicotinamide N-methyltransferase inhibition is shown to increases adipose SAM and NAD(+) levels and upregulate ornithine decarboxylase and spermidine-spermine N(1)-acetyltransferase activity and expression. (Ref.: Kraus, D et al. (2014). Nature. 508 (7495); 258-62).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nicotinamide N-methyltransferase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: 1μg/mL from a representative lot detected Nicotinamide N-methyltransferase in 10μg of  human kidney lysate and HEK293 cell lysate. Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Nicotinamide N-methyltransferase in human liver, human kidney, and human pancreas tissues.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias EC 2.1.1.1
Gene Symbols NNMT
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the internal region of human Nicotinamide N-methyltransferase.
Molecular Weight of Antigen ∽30 kDa observed; 29.57 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects human Nicotinamide N-methyltransferase. It targets an epitope within 15 amino acids from the internal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.