Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Nicotinamide Riboside
GREENER_CHOICESDP

Catalog No. 601850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-185-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-185-0 Supplier Tocris Bioscience™ Supplier No. 6018/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

NAD+ precursor; substrate for nicotinamide riboside kinases

Nicotinamide Riboside is an NAD+ precursor. It is a substrate for nicotinamide riboside kinases (NRK1/2). Nicotinamide Riboside shows neuroprotective effects in a mouse model of type 2 diabetes and improves mitochondrial function in muscle stem cells in aged mice. The compound also corrects non-alchoholic fatty liver disease phenotype induced by NAD+ deficiency or high-fat diet in mice. Orally bioavailable.

Specifications

Chemical Name or Material 3-(Aminocarbonyl)-1-β-D-ribofuranosylpyridinium chloride
CAS 23111-00-4
Quantity 50 mg
Target Enzyme Substrates
Molecular Formula C11H15ClN2O5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.