Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ (-)-Nicotine ditartrate
GREENER_CHOICESDP

Catalog No. 354650 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-465-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 35-465-0 Supplier R&D Systems™ Supplier No. 3546/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Prototypical nAChR agonist

Nicotinic acetylcholine receptor (nAChR) agonist (Ki values are 1, > 1000, 4000 and 7130 nM at α4β2, α1β1δγ, rat α7 and human α7 respectively). Exhibits vasoconstrictive, hypertensive and prothrombotic activity in vivo.

Chemical Identifiers

CAS 65-31-6
Molecular Formula C18H26N2O12

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 462.41
Purity >99%
Molecular Weight (g/mol) 462.408
Chemical Name or Material (-)-Nicotine ditartrate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.