Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotine Bitartrate Dihydrate, MP Biomedicals™

Catalog No. icn15355483 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
250 mg
1 g

Chemical Identifiers

Molecular Formula C18H30N2O14
Molecular Weight (g/mol) 498.44
MDL Number MFCD00150899
InChI Key LDMPZNTVIGIREC-HYRPJEHINA-N
SMILES O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1

Specifications

CAS 6019-06-3
Melting Point 93°C to 95°C (literature)
Color White
pH 3.18 (10% aq. soln.) (literature)
Quantity 250 mg
Synonym nicotine ditartrate dihydrate, unii-r7m676m8yv, nicotine bitartrate usan, 2r,3r-2,3-dihydroxybutanedioic acid;3-2s-1-methylpyrrolidin-2-yl pyridine;dihydrate, nicotine bitartrate who-dd, nicotine acid tartrate dihydrate, s-3-1-methylpyrrolidin-2-yl pyridine bis 2r,3r-2,3-dihydroxysuccinate dihydrate, 2r,3r-2,3-dihydroxybutanedioic acid; 3-2s-1-methylpyrrolidin-2-yl pyridine; dihydrate, 3-2s-1-methyl-2-pyrrolidinyl pyridine, 2r,3r-2,3-dihydroxybutanedioate 1:2 , dihydrate
Solubility Information Soluble in water (50mg/mL - clear to slightly hazy, colorless to faint yellow solution) or alcohol.
IUPAC Name bis((2R,3R)-2,3-dihydroxybutanedioic acid) 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine dihydrate
Formula Weight 498.44
Physical Form Powder
Chemical Name or Material Nicotine Bitartrate Dihydrate
Show More Show Less
Recommended Storage Store at Room Temperature (15 to 30°C)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.