Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotine 95.0+%, TCI America™
SDP

Catalog No. N0079100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0079100ML 100 mL
N007925ML 25 mL
N0079500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0079100ML Supplier TCI America Supplier No. N0079100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54-11-5
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
MDL Number MFCD00006369
InChI Key SNICXCGAKADSCV-JTQLQIEISA-N
Synonym nicotine, l-nicotine, --nicotine, s-nicotine, habitrol, s-3-1-methylpyrrolidin-2-yl pyridine, nicoderm, nicotrol, s---nicotine, nicoderm cq
PubChem CID 89594
ChEBI CHEBI:17688
IUPAC Name 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
SMILES CN1CCCC1C2=CN=CC=C2

Specifications

Melting Point -80°C
pH 10.2 (8.1g/L H2O soln. 20°C)
Boiling Point 91°C
Quantity 100 mL
UN Number 1654
Formula Weight 162.24
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Nicotine
EINECSNumber (9)-0990
RTECSNumber QS5250000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.