Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinic Acetylcholine R alpha 1/CHRNA1 Antibody (mAb 192), DyLight 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP281070DL594
Description
Nicotinic Acetylcholine R alpha 1/CHRNA1 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 1/CHRNA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Nicotinic Acetylcholine R alpha 1/CHRNA1 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
CHRNA1 | |
Human muscle AChR. | |
0.1 mL | |
Neuroscience | |
1134 | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
mAb 192 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
acetylcholine receptor subunit alpha, ACHRA, ACHRD, CHNRA, cholinergic receptor, nicotinic, alpha 1 (muscle), CHRNA, CMS2A, FCCMS, nicotinic acetylcholine receptor alpha subunit, nicotinic cholinergic receptor alpha 1, nicotinic, alpha polypeptide 1 (muscle), SCCMS | |
Rat | |
Protein A purified | |
RUO | |
Primary | |
The antibody binds to human muscle AChR with a Kd of 10 pM, to mouse muscle AChR with a Kd of 50 pM, and to rat muscle AChR with a Kd of 646 nM. | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction