Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinic Acetylcholine R alpha 10/CHRNA10 Antibody (4E2D1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
| Antigen | Nicotinic Acetylcholine R alpha 10/CHRNA10 |
|---|---|
| Clone | 4E2D1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61666
![]() |
Novus Biologicals
NBP261666 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
Nicotinic Acetylcholine R alpha 10/CHRNA10 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 10/CHRNA10 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| Nicotinic Acetylcholine R alpha 10/CHRNA10 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| 57053 | |
| Purified recombinant fragment of human CHRNA10 (AA: extra 25-237) expressed in E. Coli. | |
| Primary | |
| 49.7 kDa |
| 4E2D1 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| cholinergic receptor, nicotinic, alpha 10, cholinergic receptor, nicotinic, alpha polypeptide 10, NACHR alpha-10, NACHRA10, neuronal acetylcholine receptor subunit alpha-10, Nicotinic acetylcholine receptor subunit alpha-10 | |
| CHRNA10 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title