Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinic Acetylcholine R alpha 2/CHRNA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $501.50
Specifications
Antigen | Nicotinic Acetylcholine R alpha 2/CHRNA2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17995720
![]() |
Novus Biologicals
NBP17995720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179957
![]() |
Novus Biologicals
NBP179957 |
100 μL |
Each for $501.50
|
|
|||||
Description
Nicotinic Acetylcholine R alpha 2/CHRNA2 Polyclonal specifically detects Nicotinic Acetylcholine R alpha 2/CHRNA2 in Human samples. It is validated for Western Blot.Specifications
Nicotinic Acetylcholine R alpha 2/CHRNA2 | |
Polyclonal | |
Purified | |
RUO | |
NP_000733 | |
1135 | |
Synthetic peptide directed towards the N terminal of human CHRNA2. Peptide sequence TKLSLWWLLLTPAGGEEAKRPPPRAPGDPLSSPSPTALPQGGSHTETEDR. | |
Primary | |
60 kDa |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
cholinergic receptor, nicotinic, alpha 2 (neuronal), cholinergic receptor, nicotinic, alpha polypeptide 2 (neuronal), neuronal acetylcholine receptor subunit alpha-2 | |
CHRNA2 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title