Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinic Acetylcholine R alpha 4/CHRNA4 Antibody (7B4H1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Nicotinic Acetylcholine R alpha 4/CHRNA4 |
---|---|
Clone | 7B4H1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61674
![]() |
Novus Biologicals
NBP261674 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
Nicotinic Acetylcholine R alpha 4/CHRNA4 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 4/CHRNA4 in Human samples. It is validated for Western Blot, ELISA.Specifications
Nicotinic Acetylcholine R alpha 4/CHRNA4 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
BFNC, cholinergic receptor, nicotinic, alpha 4, cholinergic receptor, nicotinic, alpha polypeptide 4, EBN, EBN1, ENFL1, FLJ95812, NACHR, NACHRA4, NACRA4, neuronal acetylcholine receptor subunit alpha-4, neuronal nicotinic acetylcholine receptor alpha-4 subunit | |
CHRNA4 | |
IgG1 | |
Protein G purified |
7B4H1 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Neuroscience | |
1137 | |
Purified recombinant fragment of human CHRNA4 (AA: extra 29-242) expressed in E. Coli. | |
Primary | |
70 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title