Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic Acetylcholine R alpha 4/CHRNA4 Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody

Supplier:  Novus Biologicals NBP100143

Catalog No. NBP100143


Only null left
Add to Cart

Description

Description

Nicotinic Acetylcholine R alpha 4/CHRNA4 Polyclonal specifically detects Nicotinic Acetylcholine R alpha 4/CHRNA4 in Human, Rat samples. It is validated for Western Blot, Peptide ELISA, Immunoprecipitation (Negative).
Specifications

Specifications

Nicotinic Acetylcholine R alpha 4/CHRNA4
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
BFNC, cholinergic receptor, nicotinic, alpha 4, cholinergic receptor, nicotinic, alpha polypeptide 4, EBN, EBN1, ENFL1, FLJ95812, NACHR, NACHRA4, NACRA4, neuronal acetylcholine receptor subunit alpha-4, neuronal nicotinic acetylcholine receptor alpha-4 subunit
Goat
Affinity Purified
RUO
Primary
This antibody is expected NOT to cross-react with the similar alpha 2 subunit.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunoprecipitation
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000, Immunoprecipitation (Negative)
NP_000735.1
CHRNA4
Peptide with sequence C-HVETRAHAEERLLKK corresponding to internal region according to NP_000735.1.
0.1 mg
Neuroscience
1137
Human, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only