Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinic Acetylcholine R alpha 5/CHRNA5 Antibody (8F11G8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261678
Description
Nicotinic Acetylcholine R alpha 5/CHRNA5 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 5/CHRNA5 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Nicotinic Acetylcholine R alpha 5/CHRNA5 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Cholinergic receptor, neuronal nicotinic, alpha polypeptide-5, cholinergic receptor, nicotinic, alpha 5, cholinergic receptor, nicotinic, alpha polypeptide 5, LNCR2, NACHRA5, neuronal acetylcholine receptor subunit alpha-5, neuronal nicotinic acetylcholine receptor, alpha5 subunit | |
Mouse | |
53 kDa | |
0.1 mL | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
8F11G8 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
CHRNA5 | |
Purified recombinant fragment of human CHRNA5 (AA: extra 23-254) expressed in E. Coli. | |
Protein G purified | |
Signal Transduction | |
1138 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction