Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic Acetylcholine R alpha 7/CHRNA7 Antibody (7F10F1) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-200080700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61738 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61738 Supplier Novus Biologicals Supplier No. NBP261738
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nicotinic Acetylcholine R alpha 7/CHRNA7 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 7/CHRNA7 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen Nicotinic Acetylcholine R alpha 7/CHRNA7
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 7F10F1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias a7 nicotinic acetylcholine receptor, alpha 7 neuronal nicotinic acetylcholine receptor, alpha-7 nicotinic cholinergic receptor subunit, cholinergic receptor, nicotinic, alpha 7, cholinergic receptor, nicotinic, alpha polypeptide 7, CHRNA7-2, NACHRA7, neuronal acetylcholine receptor protein, alpha-7 chain, neuronal acetylcholine receptor subunit alpha-7
Gene Symbols CHRNA7
Host Species Mouse
Immunogen Purified recombinant fragment of human CHRNA7 (AA: extra 52-259) expressed in E. Coli.
Molecular Weight of Antigen 56.4 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1139
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.