Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinic Acetylcholine Receptor gamma Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179952
Description
Nicotinic Acetylcholine Receptor gamma Polyclonal specifically detects Nicotinic Acetylcholine Receptor gamma in Human samples. It is validated for Western Blot.Specifications
Nicotinic Acetylcholine Receptor gamma | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
acetylcholine receptor subunit gamma, acetylcholine receptor, muscle, gamma subunit, ACHRG, cholinergic receptor, nicotinic, gamma, nicotinic, gamma polypeptide | |
Rabbit | |
56 kDa | |
100 μL | |
Primary | |
Mouse: 85%; Rat: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_005190 | |
CHRNG | |
Synthetic peptide directed towards the N terminal of human CHRNG. Peptide sequence NYDPNLRPAERDSDVVNVSLKLTLTNLISLNEREEALTTNVWIEMQWCDY. | |
Affinity purified | |
RUO | |
1146 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction