Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic Acid Hydrazide 98.0+%, TCI America™
SDP

Catalog No. N0087100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0087100G 100 g
N008725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0087100G Supplier TCI America Supplier No. N0087100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 553-53-7
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006383
InChI Key KFUSANSHCADHNJ-UHFFFAOYSA-N
Synonym nicotinohydrazide, nicotinic acid hydrazide, nicotinic hydrazide, niazid, nicotinylhydrazide, nicotinoyl hydrazine, 3-pyridoyl hydrazine, 3-pyridylcarbonylhydrazine, nicotinoyl hydrazide, hydrazine, nicotinoyl
PubChem CID 11112
IUPAC Name pyridine-3-carbohydrazide
SMILES NNC(=O)C1=CC=CN=C1

Specifications

Melting Point 162°C
Color White
Quantity 100 g
Formula Weight 137.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nicotinic Acid Hydrazide
RTECSNumber QT1750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.