Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic acid N-oxide, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
Plastic bottle
2398-81-4
C6H5NO3
139.11
MFCD00006201
FJCFFCXMEXZEIM-UHFFFAOYSA-N
nicotinic acid n-oxide,oxiniacic acid,nicotinic acid 1-oxide,3-pyridinecarboxylic acid, 1-oxide,nicotinic acid oxide,n-hydroxynicotinic acid,3-carboxypyridin-1-ium-1-olate,3-carboxypyridine n-oxide,3-carboxypyridine 1-oxide,3-pyridinecarboxylic acid oxide
16976
1-oxidopyridin-1-ium-3-carboxylic acid
C1=CC(=C[N+](=C1)[O-])C(=O)O
2398-81-4
C6H5NO3
139.11
MFCD00006201
FJCFFCXMEXZEIM-UHFFFAOYSA-N
nicotinic acid n-oxide,oxiniacic acid,nicotinic acid 1-oxide,3-pyridinecarboxylic acid, 1-oxide,nicotinic acid oxide,n-hydroxynicotinic acid,3-carboxypyridin-1-ium-1-olate,3-carboxypyridine n-oxide,3-carboxypyridine 1-oxide,3-pyridinecarboxylic acid oxide
16976
1-oxidopyridin-1-ium-3-carboxylic acid
C1=CC(=C[N+](=C1)[O-])C(=O)O
Melting Point 260.0°C to 262.0°C
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Merck Index 15, 7040
Formula Weight 139.11
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material Nicotinic acid N-oxide
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning