Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinoyl chloride hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
20260-53-1
C6H4ClNO·HCl
178.02
MFCD00012820
MSYBLBLAMDYKKZ-UHFFFAOYSA-N
nicotinoyl chloride hydrochloride,pyridine-3-carbonyl chloride hydrochloride,nicotinoyl chloride, hydrochloride,3-pyridinecarbonyl chloride, hydrochloride,3-pyridinecarbonyl chloride hydrochloride,nicotinic acid chloride hydrochloride,nicotinoylchloride hydrochloride,3-pyridinecarbonyl chloride, hydrochloride 1:1,nicotinyl chloride hydrochloride,nicotinoyl chloride hcl
88438
pyridine-3-carbonyl chloride;hydrochloride
C1=CC(=CN=C1)C(=O)Cl.Cl
20260-53-1
C6H4ClNO·HCl
178.02
MFCD00012820
MSYBLBLAMDYKKZ-UHFFFAOYSA-N
nicotinoyl chloride hydrochloride,pyridine-3-carbonyl chloride hydrochloride,nicotinoyl chloride, hydrochloride,3-pyridinecarbonyl chloride, hydrochloride,3-pyridinecarbonyl chloride hydrochloride,nicotinic acid chloride hydrochloride,nicotinoylchloride hydrochloride,3-pyridinecarbonyl chloride, hydrochloride 1:1,nicotinyl chloride hydrochloride,nicotinoyl chloride hcl
88438
pyridine-3-carbonyl chloride;hydrochloride
C1=CC(=CN=C1)C(=O)Cl.Cl
Color White
Quantity 100g
Packaging Glass bottle
Melting Point 151.0°C to 156.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: decomposes
Formula Weight 178.02
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Nicotinoyl chloride hydrochloride, 97%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 243-648-1

TSCA : TSCA