Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nidogen-1/Entactin Antibody (2R33) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019295 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019295 0.025 mg
NBP222037 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019295 Supplier Novus Biologicals Supplier No. NBP2220370.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nidogen-1/Entactin Monoclonal specifically detects Nidogen-1/Entactin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Nidogen-1/Entactin
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA
Classification Monoclonal
Clone 2R33
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:2000, Immunohistochemistry 1:50-1:250, Immunohistochemistry-Paraffin 1:50 - 1:250
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. NP_002499
Gene Alias enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1
Gene Symbols NID1
Host Species Mouse
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant Nidogen-1/Entactin. Mouse anti human Nidogen-1/Entacin NP_002499.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4811
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.