Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nidogen-1/Entactin Antibody (ELM1), Janelia Fluor™ 646, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP197701JF646
Description
Nidogen-1/Entactin Monoclonal specifically detects Nidogen-1/Entactin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Nidogen-1/Entactin | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NID1 | |
Clone ELM1 is a rat monoclonal IgG2a antibody derived by fusion of X63 Ag 8.653 mouse myeloma cells with spleen cells from a Fisher rat immunized with a partially purified preparation of laminin from the EHS mouse tumor. | |
0.1 mL | |
4811 | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
ELM1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1 | |
Rat | |
Protein A or G purified | |
Primary | |
ELM1 reacts with entactin. | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction