Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nidogen-1/Entactin Rat anti-Mouse, Guinea Pig, Clone: ELM1, Novus Biologicals™

Rat Monoclonal Antibody
$268.00
Specifications
| Antigen | Nidogen-1/Entactin |
|---|---|
| Clone | ELM1 |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA 2-4 ug/ml |
| Applications | ELISA |
Description
Nidogen-1/Entactin Monoclonal specifically detects Nidogen-1/Entactin in Mouse, Guinea Pig samples. It is validated for ELISA.Specifications
| Nidogen-1/Entactin | |
| 0.2 mg/ml | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse, Guinea Pig | |
| enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1 | |
| Murine EHS laminin preparation | |
| Primary | |
| Store at 4°C. |
| ELM1 | |
| ELISA 2-4 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 4811 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title