Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nidogen 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547482
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547482 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547482 Supplier Invitrogen™ Supplier No. PA547482
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human Nidogen-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Basement membranes, which are composed of type IV collagens, laminins, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis. Basement membranes, which are composed of type IV collagens, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nidogen 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene NID2
Gene Accession No. Q14112
Gene Alias AW547149; entactin 2; entactin-2; LOW QUALITY PROTEIN: nidogen-2; Ly111; Nid2; NID-2; Nidogen; nidogen 2; nidogen 2 (osteonidogen); nidogen-2; nidogen-2; LOW QUALITY PROTEIN: nidogen-2; osteonidogen
Gene Symbols NID2
Host Species Goat
Immunogen CHO-derived recombinant human Nidogen-2 Leu31-Lys1375 (Gly832Ala).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22795
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.