Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nidogen G1 Domain Monoclonal Antibody (JF3)
GREENER_CHOICE

Catalog No. PIMA517019
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517019 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517019 Supplier Invitrogen™ Supplier No. MA517019
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MA5-17019 recognizes native and denatured mouse and human nidogen, and blocks binding of nidogen to laminin. MA5-17019 has been successfully used in ELISA and Western blot applications. The MA5-17019 immunogen is a recombinant fragment corresponding to the G1Link region of mouse nidogen produced in Fisher rat plus YB2/0 cells.

Basement membrane components play an important role in cell adhesion, locomotion, differentiation and embryonic compartmentalization. Although the precise composition and structure of basal laminae varies from tissue to tissue, as well as within the same tissue at different developmental stages, most mature basal laminae contain type IV collagen, laminins, heparan sulphate proteoglycans and entactin (or nidogen). Entactin is a sulfated glycoprotein that is widely distributed in basement membranes. It is tightly associated with laminin and also binds to collagen IV. Entactin probably has a role in cell-extracellular matrix interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nidogen G1 Domain
Applications ELISA, Inhibition Assays, Western Blot
Classification Monoclonal
Clone JF3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 2M acetic acid, 2M tris with 0.1% sodium azide
Gene NID1
Gene Accession No. P10493, P14543
Gene Alias A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; NID; NID 1; Nid1; NID-1; Nidogen (enactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1
Gene Symbols NID1
Host Species Rat
Immunogen Recombinant fragment corresponding to the G1Link region of mouse nidogen produced in Fisher rat plus YB2/0 cells.
Purification Method Protein G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18073, 4811
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.