Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nidogen G3 Domain Monoclonal Antibody (JF2)
Nidogen G3 Domain Monoclonal Antibody (JF2)
Supplier: Thermo Scientific MA517021
Description

Specifications
Nidogen G3 Domain | |
Monoclonal | |
1 mg/mL | |
0.5M acetic acid, 2M tris with 0.1% sodium azide | |
P10493, P14543 | |
NID1 | |
Recombinant fragment corresponding to the G3 region of mouse nidogen produced in Fisher rat plus YB2/0 cells. | |
50 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
ELISA, Inhibition Assays, Western Blot | |
JF2 | |
Unconjugated | |
NID1 | |
A630025O17; enactin; Ent; Ent1; Entactin; entactin 1; entactin-1; NID; NID 1; Nid1; NID-1; Nidogen (enactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1 | |
Rat | |
Protein G | |
RUO | |
18073, 4811 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction