Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nidogen Monoclonal Antibody (302117)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA523911

Catalog No. PIMA523911


Only null left
Add to Cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombinant human Nidogen-2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nidogen
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P14543
NID1
Mouse myeloma cell line NS0-derived recombinant human Nidogen-1/Entactin Leu29-Lys1114 (Gln1113Arg).
100 μg
Primary
Human
Antibody
IgG1
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
302117
Unconjugated
NID1
A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;
Mouse
Protein A/G
RUO
4811
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.