Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nidogen Monoclonal Antibody (ELM1)

Rat Monoclonal Antibody

Supplier:  Invitrogen™ MA106501

Catalog No. MA106501


Only null left
Add to Cart

Description

Description

MA1-06501 detects entactin in human samples. MA1-06501 has sucessfully been used in Western blotting, immunosytochemistry and immunohistochemistry. By Western blotting, it detects a ~150 kDa protein representing entactin. The MA1-06501 immunogen is partially purified preparation of laminin from the EHS mouse tumor. Store at 4ºC or in small aliquots at -20ºC.

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nidogen
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P10493
NID1
Partially purified preparation of laminin from the EHS mouse tumor.
100 μg
Primary
Mouse
Antibody
IgG2a
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
ELM1
Unconjugated
NID1
A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;
Rat
Purified
RUO
18073
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.