Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nidogen Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599666 Supplier Invitrogen™ Supplier No. PA599666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Nidogen.

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nidogen
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NID1
Gene Accession No. P14543
Gene Alias A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;
Gene Symbols NID1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NID1(Accession P14543), corresponding to amino acid residues I980-H1030.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4811
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.