Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niemann Pick C1 Rabbit anti-Human, Mouse, Rat, FITC, Polyclonal, FabGennix

Catalog No. 502207986
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7986 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7986 Supplier Fabgennix Inc Supplier No. NPC1FITC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Niemann Pick C1 Polyclonal antibody specifically detects Niemann Pick C1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Niemann Pick C1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate FITC
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene NPC1
Gene Accession No. O15118, O35604
Gene Alias A430089E03Rik; C85354; Cdig2; D18Ertd139e; D18Ertd723e; FLJ98532; I79_014078; lcsd; Niemann Pick type C1; Niemann-Pick C disease protein; niemann-Pick C1 protein; Niemann-Pick disease, type C1; Niemann-Pick type C1; Niemann-Pick type C1 protein; nmf164; NPC; NPC intracellular cholesterol transporter 1; NPC1; sphingomyelinosis; spm; truncated Niemann-Pick C1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1228-1278 on human Niemann-Pick C1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18145, 266732, 4864
Target Species Human, Mouse, Rat
Content And Storage -20° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.