Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIF1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002578JF646
Description
NIF1 Polyclonal antibody specifically detects NIF1 in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
NIF1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
DKFZp586G1122, Hzf, HZFzinc finger protein 385, RZFHematopoietic zinc finger protein, ZFP385, zinc finger protein 385A, ZNF385Retinal zinc finger protein | |
The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human NRC-Interacting Factor 1 using the numbering given in entry NP_071378.1 (GeneID 63925). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
63925 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction