Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIF3L1 Monoclonal Antibody (OTI1F6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503679S


Description
NIF3L1 Monoclonal antibody specifically detects NIF3L1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
NIF3L1 | |
Monoclonal | |
0.85 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9H4Y5 | |
Mouse | |
Affinity chromatography | |
RUO | |
119391 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1F6 | |
Unconjugated | |
Gsto2 | |
1110030G24Rik; ALS2CR1; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 1; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 1 protein; CALS-7; GTP cyclohydrolase I; MDS015; My018; Ngg1 interacting factor 3 homolog-like 1; NGG1 interacting factor 3 like 1; Ngg1 interacting factor 3-like 1; Ngg1 interacting factor 3-like 1 (S. pombe); NIF3 (Ngg1 interacting factor 3, S.pombe homolog)-like 1; NIF3 NGG1 interacting factor 3-like 1; NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae); Nif3l1; NIF3-like protein 1; OTTHUMP00000206190; OTTHUMP00000206191; OTTHUMP00000206192; putative GTP cyclohydrolase 1 type 2 Nif3l1 | |
Full length human recombinant protein of human GSTO2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction